Anti-SYCP3 Rabbit Polyclonal Antibody (DyLight® 549)
ABNOPAB15246
: Abnova
- Antigen name:Synaptonemal Complex Protein 3
- Antigen symbol:SYCP3
- Clonality:Polyclonal
- Conjugation:DyLight® 549
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Mouse
- Cross adsorption:No
- Format:Liquid
- Gene ID:20962
- Antigen synonyms:SPGF4|COR1|SCP3
- Storage buffer:50 mM sodium borate
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:A synthetic peptide corresponding to C-terminus of mouse Sycp3.
- Size:100 μl
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic peptide of Sycp3.
Recommended Dilutions: Immunofluorescence: 1:100-1:750; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:750
Type:
Antigen: SYCP3
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 549
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse