Anti-KLK3 Mouse Monoclonal Antibody [clone: B732M]
Catalog # ABNOMAB6733
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Kallikrein-related peptidase 3
- Antigen symbol:KLK3
- Clonality:Monoclonal
- Clone:B732M
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:354
- Antigen synonyms:PSA|APS|hK3|KLK2A1
- Storage buffer:PBS, pH 7,4 (0,05% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at– 20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Purified PSA.
- Pk:1 mg
Specifications
About this item
Mouse monoclonal antibody raised against PSA (free).
Type: Primary
Antigen: KLK3
Clonality: Monoclonal
Clone: B732M
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human