Anti-MRPS26 Rabbit Polyclonal Antibody
ABNOPAB24417
: Abnova
- Antibody type:Primary
- Antigen name:mitochondrial ribosomal protein S26
- Antigen symbol:MRPS26
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:64949
- Antigen synonyms:NY-BR-87|MRPS13|GI008|RPMS13|dJ534B8.3|C20orf193|MRP-S13|MRP-S26
- Storage buffer:PBS, pH 7,5 (40% glycerol, 0,02% sodium azide)
- Sequence:EVLQLQEEVKNFITRENLEARVEAALDSRKNYNWAITREGLVVRPQRRDS
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids of human MRPS26.
- Purification:Antigen affinity purification
- Size:100 μl
- Pk:100 µl
Rabbit polyclonal antibody raised against recombinant MRPS26.
Recommended Dilutions: Immunohistochemistry: 1:200-1:500; Western Blot: 1:250-1:500
Type: Primary
Antigen: MRPS26
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human