Anti-MEX3D Rabbit Polyclonal Antibody
Catalog # ABNOPAB16558
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Mex-3 RNA binding family member D
- Antigen symbol:MEX3D
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:399664
- Antigen synonyms:RNF193|Rkhd1|RNA-binding protein MEX3D|KIAA2031|MEX3|RING finger and KH domain-containing protein 1|MEX-3D|TINO|OK/SW-cl.4|RKHD1
- Storage buffer:PBS (0,02% sodium azide)
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to internal region 15 amino acids of human MEX3D.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of MEX3D.
Recommended Dilutions: Western Blot: 1-2 ?g/ml
Type: Primary
Antigen: MEX3D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human