Anti-VPS25 Rabbit Polyclonal Antibody
ABNOPAB5630
: Abnova
- Antibody type:Primary
- Antigen name:Vacuolar Protein Sorting 25
- Antigen symbol:VPS25
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:84313
- Antigen synonyms:EAP20|DERP9|FAP20
- Storage buffer:PBS, pH 7,2 (50% glycerol, 0,01% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to N-terminus of human VPS25.
- Size:100 μg
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic peptide of VPS25.
Type: Primary
Antigen: VPS25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human