Anti-ADAMTS3 Rabbit Polyclonal Antibody
ABNOPAB18819
: Abnova
- Antibody type:Primary
- Antigen name:ADAM Metallopeptidase With Thrombospondin Type 1 Motif, 3
- Antigen symbol:ADAMTS3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:9508
- Antigen synonyms:ADAMTS-4|KIAA0366
- Storage buffer:buffer containing 0,02% sodium azide
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to 16 amino acids at C-terminus of human ADAMTS3.
- Purification:Protein A purification
- Size:100 μg
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic peptide of ADAMTS3.
Recommended Dilutions: ELISA: 1:5000-1:20000; Western Blot: 1:200-1:500
Type: Primary
Antigen: ADAMTS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human