Order Entry
Ireland
ContactUsLinkComponent
Anti-ZBTB7A Rabbit Polyclonal Antibody
Anti-ZBTB7A Rabbit Polyclonal Antibody
  ABNOPAB19399
 :  Abnova
Anti-ZBTB7A Rabbit Polyclonal Antibody
  ABNOPAB19399
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Zinc Finger And BTB Domain Containing 7A
  • Antigen symbol:
    ZBTB7A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    51341
  • Antigen synonyms:
    pokemon|ZNF857A|FBI-1|LRF|ZBTB7|FBI1
  • Storage buffer:
    PBS (0,02% sodium azide)
  • Storage temperature:
    Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to 16 amino acids near N-terminus of human ZBTB7A.
  • Purification:
    Peptide affinity purification
  • Size:
    100 μg
  • Pk:
    100 µg

 

 

Rabbit polyclonal antibody raised against synthetic peptide of ZBTB7A.

Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunohistochemistry: 2.5 ?g/ml

Type: Primary
Antigen: ZBTB7A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse