Anti-CDCA4 Mouse Monoclonal Antibody [clone: 04sd]
ABNOMAB8874
: Abnova
- Antibody type:Primary
- Antigen name:Cell division cycle associated 4
- Antigen symbol:CDCA4
- Clonality:Monoclonal
- Clone:04sd
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:55038
- Antigen synonyms:HEPP|SEI-3/HEPP
- Storage buffer:citrate-Tris-HCl, pH7.0 (0,02% Procl300)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length of human CDCA4.
- Purification:Affinity purification
- Size:100 μl
- Pk:100 µl
Mouse monoclonal antibody raised against full length recombinant CDCA4.
Type: Primary
Antigen: CDCA4
Clonality: Monoclonal
Clone: 04sd
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human