Anti-BTBD6 Rabbit Polyclonal Antibody
Catalog # ABNOPAB17432
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:BTB (POZ) Domain Containing 6
- Antigen symbol:BTBD6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Cross adsorption:No
- Format:Liquid
- Gene ID:90135
- Antigen synonyms:BDPL
- Storage buffer:PBS, pH 7,4 (150mM NaCl, 0,02% sodium azide, 50% glycerol)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to internal of human BTBD6.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of BTBD6.
Recommended Dilutions: Immunohistochemistry: 1:50-1:100; ELISA: 1:12000
Type: Primary
Antigen: BTBD6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse