Anti-COL4A3BP Goat Polyclonal Antibody
ABNOPAB7393
: Abnova
- Antibody type:Primary
- Antigen name:Collagen, type IV, alpha 3 (Goodpasture antigen) binding protein
- Antigen symbol:COL4A3BP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:10087
- Antigen synonyms:STARD11|CERT|CERTL|GPBP|MRD34
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:DHDSAPLNNRC
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human COL4A3BP.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µl
Goat polyclonal antibody raised against synthetic peptide of COL4A3BP.
Recommended Dilutions: ELISA: 1:32000; Western Blot: 0.5-1.5 ?g/ml
Type: Primary
Antigen: COL4A3BP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse