Order Entry
Ireland
ContactUsLinkComponent
 
Anti-NANOG Rabbit Polyclonal Antibody
  ABNOPAB19186
 :  Abnova
Anti-NANOG Rabbit Polyclonal Antibody
  ABNOPAB19186
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Homeobox protein NANOG
  • Antigen symbol:
    NANOG
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    79923
  • Antigen synonyms:
    ENK|ecat4|2410002E02Rik
  • Storage buffer:
    PBS (0,02% sodium azide)
  • Storage temperature:
    Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to 19 amino acids near center region of human NANOG.
  • Purification:
    Affinity purification
  • Size:
    100 μg
  • Pk:
    100 µg

 

 

Rabbit polyclonal antibody rasied against synthetic peptide of NANOG.

Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: NANOG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat