Anti-DYKDDDDK tag Polyclonal Antibody
Catalog # ABNOPAB7705
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:DYKDDDDK tag
- Antigen symbol:DYKDDDDK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS (without Mg(2+) and Ca(2+)), 150 mM NaCl, pH 7,4 (50% glycerol, 0,02% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to DYKDDDDK tag.
- Purification:Immunoaffinity purification
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptides of DYKDDDDK tag.
Recommended Dilutions: Western Blot: 1:500-1:1000
Type: Primary
Antigen: DYKDDDDK tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: