Anti-DAP12 Polyclonal Antibody
Catalog # ABNOPAB9853
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:TYRO protein tyrosine kinase-binding protein
- Antigen symbol:DAP12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ImmunoPrecipitation:Yes
- Reactivity:Human,Mouse
- Cross adsorption:No
- Format:Liquid
- Antigen synonyms:DAP 12|TYROBP|TYRO protein tyrosine kinase binding protein|DAP12|DAP-12
- Storage buffer:PBS (0,08% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to DAP12.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against a recombinant DAP12.
Recommended Dilutions: Immunoprecipitation: 2-4 ?g/mg)
Type: Primary
Antigen: DAP12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Human, Mouse