Order Entry
Ireland
ContactUsLinkComponent
Anti-CD8a Mouse Monoclonal Antibody (violetFluor™ 450) [clone: Hit8a]
Anti-CD8a Mouse Monoclonal Antibody (violetFluor™ 450) [clone: Hit8a]
Supplier:  Tonbo Biosciences
Certificates
Anti-CD8a Mouse Monoclonal Antibody (violetFluor™ 450) [clone: Hit8a]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 75-0089-T100
  • 75-0089-T025
  • Antigen symbol:
  • CD8A
  • CD8A
  • Antigen name:
  • CD8 antigen, alpha chain
  • CD8 antigen, alpha chain
  • Antigen synonyms:
  • p32|Lyt-66|BB154331|Ly-2|Leu2|Ly-35|MAL|CD8|Ly-B
  • p32|Lyt-66|BB154331|Ly-2|Leu2|Ly-35|MAL|CD8|Ly-B
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Clone:
  • Hit8a
  • Hit8a
  • Reactivity:
  • Rhesus monkey, Chimpanzee, Human
  • Rhesus monkey, Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 925
  • 925
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • 2...8 °C, protected from light
  • 2...8 °C, protected from light
  • Tested applications:
  • Flow Cytometry
  • Flow Cytometry

Specifications

Related Information

Frequently Bought Together