Order Entry
Ireland
ContactUsLinkComponent
Anti-INSR Rabbit Polyclonal Antibody
Anti-INSR Rabbit Polyclonal Antibody
Catalog # ABGEAP20338A
Supplier:  Abgent
Anti-INSR Rabbit Polyclonal Antibody
Catalog # ABGEAP20338A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    insulin receptor
  • Antigen symbol:
    INSR
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Gene ID:
    3643
  • Antigen synonyms:
    HHF5|CD220
  • Amino acid number:
    826 to 854
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    156.333 kDa
  • Sequence:
    SARTMPEAKADDIVC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.77
  • Shipping temperature:
    4 °C
  • Immunogen:
    This INSR antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 826-854 amino acids from the N-terminal region of human INSR.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: INSR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human