Order Entry
Ireland
ContactUsLinkComponent
 
Anti-VAPB Rabbit Polyclonal Antibody
  ABGEAP20627C
 :  Abgent
Anti-VAPB Rabbit Polyclonal Antibody
  ABGEAP20627C
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    VAMP (vesicle-associated membrane Protein)-associated protein B
  • Antigen symbol:
    VAPB
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Epitope:
    internal
  • Gene ID:
    9217
  • Antigen synonyms:
    ALS8|VAP-B|VAMP-B
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    27.228 kDa
  • Sequence:
    PAENDKPHDVEINK
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    1.100
  • Shipping temperature:
    4 °C
  • Immunogen:
    This VAPB antibody is generated from a rabbit immunized with a KLH conjugated synthetic peptide between 126-139 amino acids from the Central region of human VAPB.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: VAPB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human, Rat