Anti-HIGD1C Rabbit Polyclonal Antibody
Catalog # ABGEAP19281B
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:HIG1 hypoxia inducible domain family, member 1C
- Antigen symbol:HIGD1C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Gene ID:613227
- Antigen synonyms:GM921
- Amino acid number:69 to 97
- Storage buffer:pH 7,4
- Molecular weight:11.079 kDa
- Sequence:CKDYIRPRFFSESKK
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.65
- Shipping temperature:4 °C
- Immunogen:This HIGD1C antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 69-97 amino acids from the C-terminal region of human HIGD1C.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: HIGD1C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Mouse