Order Entry
Ireland
ContactUsLinkComponent
Anti-ARC Rabbit Polyclonal Antibody
Anti-ARC Rabbit Polyclonal Antibody
Catalog # CAYM160737-1
Supplier:  Cayman Chemical
Anti-ARC Rabbit Polyclonal Antibody
Catalog # CAYM160737-1
Supplier:  Cayman Chemical

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    activity-regulated cytoskeleton-associated protein
  • Antigen symbol:
    ARC
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Amino acid number:
    191 to 208
  • Storage buffer:
    PBS, pH 7,2, containing 0,02% sodium azide
  • Sequence:
    PDPEPEPDFEERDESEDS
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    human ARC amino acids 191-208
  • Purification:
    purified by affinity chromatography
  • Size:
    1 Ea
  • Pk:
    1

Specifications

About this item

Type: Primary
Antigen: ARC
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse