Anti-HMGB1 Rabbit Polyclonal Antibody
Catalog # CAYM11515-1
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen symbol:HMGB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:New world monkey,Chicken,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Amino acid number:100 to 150
- Storage buffer:PBS containing 0,05% BSA and 0,05% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:human HMGB1 amino acids 100-150
- Purification:purified by Protein A chromatography
- Size:1 Ea
- Pk:1
Specifications
About this item
Type: Primary
Antigen: HMGB1
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Chicken, Human, Mouse, New World Monkey, Rat