Anti-SMAD6 Rabbit Polyclonal Antibody
CAYM10839-100
- Antibody type:Primary
- Antigen symbol:SMAD family member 6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:New world monkey,Sheep,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Amino acid number:85 to 99, 372 to 388
- Storage buffer:PBS containing 0,05% BSA and 0,05% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:synthetic peptides corresponding to amino acids 85-99 and amino acids 372-388 of human SMAD6
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: SMAD6
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, New World Monkey, Mouse, Rat, Sheep