Anti-Cytokeratin Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: C-11]
Catalog # CAYM10478-1
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen name:Cytokeratin
- Clonality:Monoclonal
- Clone:C-11
- Conjugation:PE (Phycoerythrin)
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Storage buffer:100 mM sodium phosphate, pH 7,4, containing 50 mM sucrose, 150 mM sodium chloride, 0,1% BSA, and 2 mM sodium azide
- Storage temperature:+2°C to +8 °C
- Shipping temperature:2 °C to 8 °C
- Purification:purified
- Size:1 Ea
- Pk:1
Specifications
About this item
Type: Primary
Antigen: Cytokeratin
Clonality: Monoclonal
Clone: C-11
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human