Anti-PSD95 Rabbit Polyclonal Antibody
Catalog # CAYM10009506-1
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen symbol:PSD95
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Epitope:near N-terminus
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:peptide corresponding to amino acid residues from the N-terminal region of rat PSD95
- Purification:purified by affinity chromatography
- Size:1 Ea
- Pk:1
Specifications
About this item
Type: Primary
Antigen: PSD95
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: near N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Mouse