Order Entry
Ireland
ContactUsLinkComponent
Anti-SPHK1 Rabbit Polyclonal Antibody
Anti-SPHK1 Rabbit Polyclonal Antibody
Catalog # CAYM10006822-1
Supplier:  Cayman Chemical
Anti-SPHK1 Rabbit Polyclonal Antibody
Catalog # CAYM10006822-1
Supplier:  Cayman Chemical

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    SPHK1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Amino acid number:
    264 to 274
  • Sequence:
    DLESEKYRRLG
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    human SPHK 1 amino acids 264-274
  • Purification:
    purified by affinity chromatography
  • Size:
    1 Ea
  • Pk:
    1

Specifications

About this item

Type: Primary
Antigen: SPHK1
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse