Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # CAYM10004301-1
Supplier: Cayman Chemical
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Goat
- Reactivity:Rabbit
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Size:1 Ea
- Pk:1
Specifications
About this item
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish PE (Phycoerythrin)roxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit