Anti-OPTN Rabbit Polyclonal Antibody
Catalog # CAYM100002-500
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen symbol:OPTN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Cross adsorption:No
- Amino acid number:115 to 130
- Storage buffer:TBS, pH 7,4, containing 50% glycerol, 0,5 mg/ml BSA, and 0,02% sodium azide
- Molecular weight:74 kDa
- Sequence:KGKSERSSEDPTDDSR
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:human optineurin amino acids 115-130
- Purification:purified by affinity chromatography
- Size:500 µl
- Pk:500 µl
Specifications
About this item
Type: Primary
Antigen: OPTN
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human