Anti-SMAD6 Rabbit Polyclonal Antibody
Catalog # ROCK200-401-I15
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen symbol:SMAD family member 6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Cross adsorption:No
- Gene ID:4091
- Antigen synonyms:MADH7|AOVD2|HsT17432|MADH6
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,05% (w/v) Sodium Azide
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:SMAD6 Antibody was produced from whole rabbit serum prepared by repeated immunizations with a peptide corresponding to two internal amino acid sequences in the human SMAD6 protein.
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Anti-SMAD6 antibody is suitable for use in IHC and WB. Expect a band approximately 53kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: Western Blot: 1-3 µg/ml; Immunochemistry: 5-10 µg/ml
Type: Primary
Antigen: SMAD6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Human