Anti-IgG3 Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # ROCK610-102-043
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG3
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Mouse
- Western blot:Yes
- ChIP:Yes
- Tested applications:Fluoresence ELISA
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Anti-Mouse IgG3 secondary antibody is suitable for Flow Cytometry, immunochemistry and Immunofluorescence Microscopy, ELISA, western blotting as well as other anti IgG2b antibody based assays.
Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:200 - 1:500; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: IgG3 γ
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse