Order Entry
Ireland
Orders LinkContactUsLinkComponent
Anti-MAP2K1 Rabbit Polyclonal Antibody

Anti-MAP2K1 Rabbit Polyclonal Antibody

100 µl
Catalog # ROCK600-401-D73
Anti-MAP2K1 Rabbit Polyclonal Antibody
Catalog # ROCK600-401-D73
Anti-MAP2K1 Rabbit Polyclonal Antibody
Anti-MAP2K1 Rabbit Polyclonal Antibody

100 µl

Price
1 000,00 €
Log in for availability
1 * 100 µl
Product Details & Documents

Anti-MEK1 pT386 (Rabbit) antibody is suitable for use in Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 45 kDa in size corresponding to the MEK1 Protein phosphorylated at Thr386.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: MAP2K1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human

Specifications
  • Size:
    100 µl
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Modification:
    phospho-Thr386
  • Purification:
    affinity-purified
  • Antibody type:
    Primary
  • Form:
    Liquid
  • Isotype:
    IgG
  • Storage temperature:
    –20 °C
  • Host:
    Rabbit
  • Cross adsorption:
    No
  • Shipping temperature:
    Dry Ice
  • Antigen name:
    Mitogen-activated protein kinase kinase 1
  • Cat. No.:
    ROCK600-401-D73
  • Reactivity:
    Human|Rat|Mouse
  • Format:
    Antibody fraction
  • Gene ID:
    5604
  • Antigen synonyms:
    MEK1|MAPKK1|CFC3|PRKMK1|MKK1
  • Immunogen:
    Anti-MEK1 pT386 Antibody was produced in rabbit by repeated immunizations with synthetic phospho-peptide corresponding to amino acid residues surrounding Thr 386 conjugated to KLH.
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    MAP2K1
  • Storage buffer:
    0,01 M HEPES, 0,15 M Sodium Chloride, pH 7,50,1 mg/ml Bovine Serum Albumin - IgG and Protease free, 50% (v/v) Glycerol
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.