Anti-YWHAB Rabbit Polyclonal Antibody
Catalog # ROCK600-401-D02
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, beta
- Antigen symbol:YWHAB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid
- Gene ID:56011
- Modification:phospho-Ser58
- Storage buffer:0,01 M HEPES, 0,15 M Sodium Chloride, pH 7,50,1 mg/ml Bovine Serum Albumin - IgG and Protease free, 50% (v/v) Glycerol
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Anti-14-3-3 pS58 antibody was produced by repeated immunizations with synthetic phospho-peptide corresponding to amino acid residues surrounding Ser58.
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Anti-14-3-3 pS58 antibody is suitable for use in ELISA, Western Blotting and IHC. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 29 kDa in size corresponding to the 14-3-3 Proteins phosphorylated at Ser58.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:1000; Immunochemistry: 1:500
Type: Primary
Antigen: YWHAB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat