Anti-GDF15 Rabbit Polyclonal Antibody
Catalog # ROCK209-401-B88S
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Growth differentiation factor 15
- Antigen symbol:GDF15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:9518
- Antigen synonyms:MIC-1|GDF-15|NAG-1|PTGFB|PDF|PLAB|MIC1
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:This Protein-A purified antibody was prepared by repeated immunizations with an MBP-tagged recombinant Protein produced in E.coli corresponding to mature human NAG-1 Protein. Cross reactivity to MBP was removed via cross-adsorption chromatography..
- Purification:Protein A purified
- Size:25 µl
- Pk:25 µl
Specifications
About this item
This Protein-A purified, MBP-cross-adsorbed antibody is suitable for ELISA and western blotting of human and mouse NAG-1 Protein. For detection of NAG-1 in human serum, a sandwich ELISA is suggested using this antibody in combination with anti-NAG-1/GDF.
Recommended Dilutions: ELISA: 1:500; Western Blot: 1:800 - 1:1500;
Restore with deionized water (or equivalent); 100 µl
Type: Primary
Antigen: GDF15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse