Anti-GST Rabbit Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
Catalog # ROCK600-402-200
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Glutathione-S-Transferase
- Antigen symbol:GST
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Mouse
- Western blot:Yes
- ChIP:Yes
- Size:100 µl
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: GST
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity: