Anti-CCL4 Rabbit Polyclonal Antibody
Catalog # ROCK214-401-B85S
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:chemokine (C-C motif) ligand 4
- Antigen symbol:CCL4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Pig
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:396668
- Antigen synonyms:MIP1b|SCY4A|MIP-1BETA
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:This Protein-A purified antibody was prepared from whole rabbit serum produced by repeated immunizations with recombinant Protein raised in yeast, corresponding to the 69 amino acids of the mature swine CCL4/MIP1beta Protein.
- Purification:Protein A purified
- Size:25 µl
- Pk:25 µl
Specifications
About this item
This Protein-A purified antibody is suitable for ELISA and Western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 7.8 kDa in size corresponding to swine CCL4 Protein by western blotting.
Recommended Dilutions: ELISA: >1:5000; Western Blot: 1:500;
Type: Primary
Antigen: CCL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pig