Anti-CCL20 Rabbit Polyclonal Antibody
Catalog # ROCK210-401-B51
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:chemokine (C-C motif) ligand 20
- Antigen symbol:CCL20
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Gene ID:20297
- Antigen synonyms:MIP3A|exodus-1|Scya20|CKb4|MIP-3[a]|ST38|MIP-3A|LARC
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2,
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:This purified antibody was prepared from whole rabbit serum produced by repeated immunizations with full length recombinant mouse MIP-3a Protein.
- Purification:Protein A purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
This purified antibody has been tested for use in neutralizations, ELISA and western blotting. Reactivity is also expected in radioimmunoassay.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:1000;
Restore with deionized water (or equivalent); 100 µl
Type: Primary
Antigen: CCL20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Mouse