Anti-GPC1 Rabbit Polyclonal Antibody
ROCK200-401-B44
- Antibody type:Primary
- Antigen name:Glypican 1
- Antigen symbol:GPC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:Internal
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:2817
- Antigen synonyms:glypican
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Anti-Glypican-1 Protein A purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region of human glypican-1 Protein.
- Purification:Protein A purified
- Size:100 µg
- Pk:100 µG
Anti-Glypican-1 Protein A purified antibody has been tested for use in ELISA, and immunochemistry, and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~61 kDa in size.
Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:500- 1:2000; Immunochemistry: 10µg/ml;
Type: Primary
Antigen: GPC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human