Order Entry
Ireland
ContactUsLinkComponent
Anti-KCNT1 Mouse Monoclonal Antibody (Unconjugated) [clone: S3-26]
Anti-KCNT1 Mouse Monoclonal Antibody (Unconjugated) [clone: S3-26]
Catalog # ROCK200-301-G46
undefined
Anti-KCNT1 Mouse Monoclonal Antibody (Unconjugated) [clone: S3-26]
Catalog # ROCK200-301-G46

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    potassium channel, sodium-activated subfamily T, member 1
  • Antigen symbol:
    KCNT1
  • Clonality:
    Monoclonal
  • Clone:
    S3-26
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    near the C-terminus
  • Cross adsorption:
    No
  • Format:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Gene ID:
    60444
  • Antigen synonyms:
    Slack|rSlo2
  • Storage buffer:
    50% (v/v) Glycerol
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Dry Ice
  • Immunogen:
    SLACK potassium channel Antibody was produced in mice by repeated immunizations raised against a fusion Protein near the c-terminus region of rat Slo2.2 (Slack).
  • Purification:
    Protein G purified
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Anti-SLACK Antibody is suitable for use in WB and IHC. Expect a band approximately ~140kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user. Antibody is provided in PBS pH 7.4.

Recommended Dilutions: Immunofluorescence: 1.0-10 µg/ml; Western Blot: 1 µg/ml; Immunochemistry: 0.1-1.0 µg/ml

Type: Primary
Antigen: KCNT1
Clonality: Monoclonal
Clone: S3-26
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat