Anti-IgG (H+L) Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
ROCK620-102-440
- Antibody type:Secondary
- Antigen symbol:IgG (H+L)
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Goat
- ImmunoFluorescence:Yes
- Reactivity:Golden syrian hamster,Armenian hamster
- ChIP:Yes
- Tested applications:Fluoresence ELISA, Fluoresence Western Blot
- Size:1 mg
- Pk:1 mg
Recommended Dilutions: FLISA: 1:10000-1:50000;
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Armenian, Golden Syrian Hamster