Order Entry
Ireland
ContactUsLinkComponent
 
Anti-GRIN2A Rabbit Polyclonal Antibody
  ROCK100-401-C86
undefined
Anti-GRIN2A Rabbit Polyclonal Antibody
  ROCK100-401-C86

 

  • Antibody type:
    Primary
  • Antigen name:
    Glutamate receptor, ionotropic, N-methyl D-aspartate 2A
  • Antigen symbol:
    GRIN2A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Cross adsorption:
    No
  • Format:
    Antiserum
  • Form:
    Liquid
  • Gene ID:
    24409
  • Antigen synonyms:
    NR2A|GluN2A|NMDAR2A
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Dry Ice
  • Immunogen:
    Anti-NMDA R2A Antibody was produced by repeated immunizations with fusion Protein from the C-terminal region of the NR2A subunit
  • Size:
    50 µl
  • Pk:
    50 µl

 

 

Anti-NMDA R2A Antibody is suitable for use in Western Blotting and IHC. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 180 kDa in size corresponding to the NR2A subuint of the NMDA receptor.

Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:500;

Type: Primary
Antigen: GRIN2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Mouse, Human, Rat