Anti-BBU94A_0149 Rabbit Polyclonal Antibody
ROCK200-401-C14
- Antibody type:Primary
- Antigen symbol:BBU94A_0149
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:B. burgdorferi b31,B. burgdorferi
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Lyophilized
- Gene ID:7106737
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:MBP-fusion Protein corresponding to Borrelia burgdorferi Flagellin Protein.
- Purification:Protein A purified
- Size:100 µg
- Pk:100 µG
This Protein-A purified antibody has been tested for use in Western blotting. Specific conditions for reactivity should be optimized by the user. Expect a band approximately 33.9 kDa in size corresponding to Borrelia burgdorferi Flagellin Protein by Western blotting.
Recommended Dilutions: ELISA: 1:6000; Western Blot: 1:1000;
Type: Primary
Antigen: BBU94A_0149
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Borrelia burgdorferi B31, Borrelia burgdorferi