Anti-H5N1 Mouse Monoclonal Antibody (Unconjugated) [clone: 15A3]
Catalog # ROCK500-301-975
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Hemagglutinin 5
- Clonality:Monoclonal
- Clone:15A3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Virus
- Western blot:Yes
- Cross adsorption:No
- Neutralisation:Yes
- Format:Ascites
- Form:Liquid (sterile filtered)
- Storage buffer:0,01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:This monoclonal antibody was produced by intraperitoneal immunization of BALB/c mice with concentrated purified virus preparation containing hemagglutinin (HA) Protein of influenza A virus (strain A/Vietnam/1203/04 (H5N1)) using the modification of the me
- Size:100 µl
- Pk:100 µl
Specifications
About this item
This monoclonal antibody can be used for hemagglutination inhibition (HI) assays to provide antigenic characterization of the influenza A viruses of the H5 HA subtype.
Recommended Dilutions: ELISA: 1:5000;
Type: Primary
Antigen: H5N1
Clonality: Monoclonal
Clone: 15A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Virus, H5N1 Virus