Anti-MHC Class 1 HLA ABC Mouse Monoclonal Antibody [clone: 5B39]
Catalog # USBIM3886-13-100
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:MHC Class 1 HLA ABC
- Clonality:Monoclonal
- Clone:5B39
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,5% saccharose, 0,2% gelatin,0,1% sodium azide.
- Storage temperature:–20 °C
- Concentration:~1 mg/ml
- Immunogen:HLA ABC
- Purification:purified by Protein A chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: MHC Class 1 HLA ABC
Clonality: Monoclonal
Clone: 5B39
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human,