Anti-RUNX3 Rabbit Polyclonal Antibody
Catalog # USBIR9915-20A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:RUNX3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,02% sodium azide, 30% glycerol.
- Storage temperature:–20 °C
- Concentration:~0,5 mg/ml
- Immunogen:Synthetic peptide corresponding to mouse RUNX3, conjugated to KLH
- Purification:purified by immunoaffinity chromatography
- Pk:40 µG