Anti-PRDX6 Rabbit Polyclonal Antibody
USBIP3352-32B
- Antibody type:Primary
- Antigen name:Peroxiredoxin 6
- Antigen symbol:PRDX6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,02% sodium azide, 30% glycerol.
- Storage temperature:–20 °C
- Concentration:~0,5 mg/ml
- Immunogen:Synthetic peptide (KLH-conjugated) of human Peroxiredoxin-6.
- Purification:purified by immunoaffinity chromatography
- Size:40 µg
- Pk:40 µG