Anti-Cofilin Rabbit Polyclonal Antibody
USBIC7506-10D
- Antibody type:Primary
- Antigen name:Cofilin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:lyophilized powder
- Storage buffer:PBS, pH 7,4, 0,02% sodium azide.
- Storage temperature:–20 °C
- Concentration:~0,5 mg/ml
- Immunogen:Synthetic peptide from amino terminal of human cofilin protein (Swiss Prot: P23528).
- Purification:purified by immunoaffinity chromatography
- Size:40 µg
- Pk:40 µG