Anti-Resistin-like Molecule, alpha Rabbit Polyclonal Antibody
Catalog # USBIR1587-04A-100
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:Resistin-like Molecule, alpha
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:lyophilized powder
- Storage temperature:–20 °C
- Concentration:~1 mg/ml
- Immunogen:Highly pure (> 98%) recombinant murine RELM alpha
- Purification:purified by immunoaffinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: >0.5µg/ml
Type: Primary
Antigen: Resistin-like Molecule, alpha
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse,