Anti-FtsZ Rabbit Polyclonal Antibody
AGRIAS10715
: Agrisera
- Antibody type:Primary
- Antigen symbol:FTSZ
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:A. thaliana
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from known bacterial sequences of FtsZ including E.coli P0A9A6
- Size:200 µl
- Pk:50 µl
Type: Primary
Antigen: FTSZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana