Anti-3-nitroY Mouse Monoclonal Antibody
Catalog # AGRIAS10706-25
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen name:3-(4-Hydroxy-3-nitrophenylacetamido) propionic acid
- Antigen symbol:3-nitroY
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:BSA-conjugated 3-(4-Hydroxy-3-nitrophenylacetamido) propionic acid
- Size:25 µl
- Pk:25 µl
Specifications
About this item
Type: Primary
Antigen: 3-nitroY
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Mouse, Rat