Order Entry
Ireland
ContactUsLinkComponent
Anti-Biotin Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Anti-Biotin Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Catalog # AGRIAS101455
Supplier:  Agrisera
Anti-Biotin Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Catalog # AGRIAS101455
Supplier:  Agrisera

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    Biotin
  • Clonality:
    Polyclonal
  • Conjugation:
    TRITC (Tetramethylrhodamine Isothiocyanate)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Cross adsorption:
    No
  • Storage temperature:
    +2°C to +8 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    Biotin
  • Size:
    1 mg
  • Pk:
    1 mg

Specifications

About this item

Type: Secondary
Antigen: Biotin
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: Goat
Isotype:
Reactivity: