Anti-Ricin Mouse Monoclonal Antibody [clone: MMH]
AGRIAS09649
: Agrisera
- Antibody type:Primary
- Antigen symbol:RICIN
- Clonality:Monoclonal
- Clone:MMH
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:A. thaliana
- Cross adsorption:No
- Storage temperature:–70 °C
- Shipping temperature:Ambient
- Immunogen:ricin, whole molecule
- Size:200 µg
- Pk:200 µG
Type: Primary
Antigen: RICIN
Clonality: Monoclonal
Clone: MMH
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: A. thaliana