Anti-Lhcb1 Rabbit Polyclonal Antibody
AGRIAS09522
: Agrisera
- Antibody type:Primary
- Antigen symbol:LHCB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Spinacia oleracea,A. thaliana,H. vulgare
- Western blot:Yes
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:BSA-conjugated synthetic peptide derived from Arabidopsis thaliana At1g29910 (Lhcb1.1), At1g29920 (Lhcb1.2), At1g29930 (Lhcb1.3, most expressed), At2g34430 (Lhcb1.4), and At2g34420 (Lhcb1.5)
- Size:100 µg
- Pk:100 µG
Type: Primary
Antigen: LHCB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, H. vulgare, Spinacia oleracea