Order Entry
Ireland
ContactUsLinkComponent
 
Anti-HSP90-1 Rabbit Polyclonal Antibody
Catalog # AGRIAS08346
Supplier:  Agrisera
Anti-HSP90-1 Rabbit Polyclonal Antibody
Catalog # AGRIAS08346
Supplier:  Agrisera

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HSP90-1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    C. reinhardtii,
    Desmodesmus subspicatus,
    Physcomitrella patens
  • Western blot:
    Yes
  • Epitope:
    full length
  • Cross adsorption:
    No
  • UniProtKB:
    P27323
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ambient
  • Immunogen:
    full length recombinant Hsp90-1 from Arabidopsis thaliana,
  • Size:
    100 µl
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: HSP90-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: full length
Host: Rabbit
Isotype:
Reactivity: Chlamydomonas reinhardtii, Desmodesmus subspicatus, Physcomitrella patens